Learn More
Rogo Sampaic™ Class B Glass Burettes
Descrizione
- With funnel
- Straight glass faucet
- Permanent red graduation marks
- Corrosion-free and resistant to thermal shock
Specifica
Specifica
| Capacità (sistema metrico) | 10 mL |
| Materiale | Borosilicate Glass |
| Graduazioni | 0.1 mL |
| Tolleranza | ±0.05 mL |
| Tipo | Buret |
| Class | Class B |