Learn More
Kimble Chase™ Borosilicate Glass Color Coded Serological Pipets
Marca: Kimble Chase™ 37034A2110
Dettagli aggiuntivi : Peso : 0.80000kg
Specifica
Borosilicate Glass | |
0.1 mL | |
±0.02 mL | |
2 mL | |
Serological Pipette |
Yes | |
6/Cs. | |
Green | |
350 mm |