Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale
Codice prodotto. 11822662
Cambia vista
Click to view available options
Capacità (sistema metrico):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Dimensione della confezione:
Pezzo
pezzi 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Capacità (sistema metrico) unitSize
11822662 1000 mL pezzi 2
10110551 250 mL pezzi 2
11892652 100 mL pezzi 2
11846164 2000 mL Pezzo
11812662 500 mL pezzi 2
11822652 50 mL pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Codice prodotto. 11822662 Fornitore Lenz Laborglasinstrumente™ N. del fornitore 04016170
  • ONLINE EXCLUSIVE PRICE
    159.00€
    124.19€ / pezzi 2
    Risparmia 34.81€
    21% Sconto
Spedizione stimata: 27-08-2026
Aggiungi al carrello
Aggiungi al carrello
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with PTFE Plug, Scale are used in liquid-liquid extractions to separate the components of a mixture into immiscible solvent phases of different densities.
  • With NS socket and PE stopper
  • with PTFE plug
  • with screwthread retaining device
  • with scale (approx. volume /ml)
  • material: borosilicate glass 3.3
Specifica
Capacità (sistema metrico) 1000 mL
Materiale Borosilicate Glass
Quantità 2Pack
Stopper Material Polyethylene