Learn More
Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale
Product Code. 11892642
Change view
Click to view available options
Capacity (Metric):
50 mL
100 mL
250 mL
500 mL
1000 mL
2000 mL
Unit Size:
Each
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11892642 250 mL Pack of 2
11886204 2000 mL Each
11812652 1000 mL Pack of 2
11882642 100 mL Pack of 2
11802652 500 mL Pack of 2
11872642 50 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 11892642 Supplier Lenz Laborglasinstrumente™ Supplier No. 04012049
  • ONLINE EXCLUSIVE PRICE
    137.00€
    72.48€ / pezzi 2
    Risparmia 64.52€
    47% Sconto
Spedizione stimata: 10-12-2026
For use in liquid-liquid extractions to separate components. Lenz Laborglasinstrumente™ Borosilicate Glass Squibb Separating Funnels with Solid Glass Plug, Scale are manufactured from durable DURAN™ glass.
  • With NS socket and PE stopper
  • Scale of approx. volume/mL
  • With screwthread retaining device
Specifications
Capacity (Metric) 250 mL
Material Borosilicate Glass
Quantity 2Pack
Stopper Material Polyethylene