Learn More
R&D Systems™ MgATP Solution
Codice prodotto. 30002141
Cambia vista
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
30002141 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30002141 Fornitore R&D Systems™ N. del fornitore B20
  • ONLINE EXCLUSIVE PRICE
    170.00€
    160.65€ / 100µL
    Risparmia 9.35€
    5% Sconto
 Articolo soggetto a restrizioni
In stock
General Ubiquitin Proteasome Pathway (UPP) Products
Pre-coupled Mg-ATP is an ideal energy source for semi-purified conjugation/degradation reactions. Supplied as a pH adjusted solution with no added buffering reagents.
Stock: 0.1M Mg2+-ATP, pH 7.0
TRUSTED_SUSTAINABILITY
Specifica
Quantità 100 μL
Requisiti di stoccaggio Store the unopened product at -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.