Learn More
Sampling Systems™ Bottle Holder
Product Code. 17992042 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1 L
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
17992042 500 mL Each
17962042 1 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17992042 Supplier Sampling Systems™ Supplier No. 5065D502
  • ONLINE EXCLUSIVE PRICE
    57.20€
    51.04€ / pezzi 1
    Risparmia 6.16€
    10% Sconto
View all promotions
Spedizione stimata: 18-12-2026
Add to basket
Add to basket
Securely holds bottles up to 88 mm in diameter on the TeleScoop. The angle can be set prior to taking the liquid samples.
  • Telescopic samplers are often used for taking surface samples from rivers, tanks and lakes
  • A wide choice of attachments is available to make the TeleScoop ideally suited for most applications
  • Attachments are interchangeable and simply click into position
Specifications
Product Type Dipper
Material HDPE
Autoclavable Not Autoclavable
Capacity (Metric) 500 mL
Disposable Reusable
Graduated No
Sterility Non-sterile