Learn More
Tocris Bioscience™ BPTU
P2Y1 allosteric antagonist
Marca: Tocris Bioscience™ 6078/50
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
BPTU is an allosteric antagonist of P2Y1 (EC50 = 0.06-0.3 μM). Non-nucleotide ligand. Binds receptor outside of the helical bundle. Blocks inhibition of spontaneous contraction of rat and mouse colon induced by electrical field stimulation, nicotine and P2Y agonists. Antithrombotic; reduces platelet aggregation.Specifica
| N-[2-[2-(1,1-Dimethylethyl)phenoxy]-3-pyridinyl]-N′-[4-(trifluoromethoxy)phenyl]urea | |
| 870544-59-5 | |
| >98% | |
| 50 mg |