Learn More
Brady™ BradyBondz™ WorkHorse™ Series Thermal Transfer Printable Label
BradyBondz™ WorkHorse™ Thermal Transfer Printable Clear Polyester (B-430) labels are designed for applications that use barcodes, alphanumerics, graphic symbols and logos and require nameplate-like quality.
Brand: Brady™ 030189
Description
- Glossy, clear labels are thermal transfer printable and feature a durable permanent acrylic adhesive
- Excellent resistance to most chemicals, oils and water
- RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC; UL approved; CSA accepted
- Service temperature of -70 to 100° F (-56 to 137° C)
Tekniske data
| Autoclavable | |
| Monochrome | |
| Permanent | |
| Gloss | |
| Polyester | |
| Non-sterile | |
| Brady 1244, Brady 1344, Brady 2461, Brady 3481, Brady 6441, Brady 300MVP Plus, Brady 200MVP Plus, Brady 300X-Plus II, Brady 360X-Plus II, Brady 600X-Plus II, Tagus T200, Tagus T300, Thermal Transfer Printer, IP300 Thermal Transfer Printer, IP600 Thermal Transfer Printer, BBP81 Label Printer | |
| 3000/Roll | |
| -70°C to +100°C | |
| 0.076 mm | |
| 3 in. | |
| 1 in. | |
| 3 in. | |
| THT-18-430-3 |
| UL Recognized, CSA Approved | |
| Thermal Transfer Printable Labels | |
| Clear | |
| Roll | |
| Thermal Transfer | |
| Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates | |
| THT-18-430-3 | |
| Rectangle | |
| 0.003 in. | |
| 1 x 3 in. | |
| 76.2 mm | |
| 25.4 mm | |
| 76.20 mm |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.