Learn More
Brannan™ Brass Slot Front Case
Product Code. 13009182
Change view
Click to view available options
Length (Metric):
405 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
13009182 405 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13009182 Supplier Brannan™ Supplier No. 47/932/0
  • ONLINE EXCLUSIVE PRICE
    8.55€
    7.82€ / pezzi 1
    Risparmia 0.73€
    8% Sconto
Spedizione stimata: 10-12-2026
Specifications
Length (Metric) 405 mm
Product Type Brass Slot Front Case
For Use With (Equipment) For thermometer