Learn More
Brady™ Breaker Blocker Component - Breaker Blocker Component - Red Blocking Bar
Product Code. 15633517 Shop All Brady Products
Change view
Click to view available options
Color:
Red
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15633517 Red Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15633517 Supplier Brady™ Supplier No. 090892
  • ONLINE EXCLUSIVE PRICE
    35.80€
    32.88€ / pezzi 5
    Risparmia 2.92€
    8% Sconto
In stock
Helps to prevent electrical risk.
  • Red bar indicates locked ‘off’
Specifications
Product Type Breaker Blocker Components
Alternate Part Number EZ 480/600 VOLT 7.6 in. RED BLOCK BAR PK5
Color Red
Description Breaker Blocker Components - 190 mm Red Blocking Bar
Quantity 5 Pk.