Learn More
Novus Biologicals™ Breast Matched Tumor and Normal Tissue Lysate
Codice prodotto. 18129417 Sfoglia Tutto Bio Techne Prodotti
Cambia vista
Click to view available options
Quantità:
2 Vials
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
18129417 2 Vials pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18129417 Fornitore Novus Biologicals™ N. del fornitore NBP247113
 Articolo soggetto a restrizioni
Spedizione stimata: 13-10-2026
Two matched 1mg/mL vials (100μg/vial) of human breast tissue lysate, one containing normal tissue and the other tumor tissue, for use in research. Lysates are guaranteed for 6 months from date of receipt.
The Breast Matched Tumor & Normal Tissue Lysate is derived from human.
TRUSTED_SUSTAINABILITY
Specifications
Tissue Breast
Quantità 2 Vials
Specie ospite Human
Content And Storage Store at −80°C. Avoid freeze-thaw cycles.