Learn More
Brilliant Stain Buffer Plus (RUO), BD Biosciences™
Codice prodotto. 15815009 Sfoglia Tutto BD Biosciences Prodotti
Cambia vista
Click to view available options
Quantità:
1000 Tests
Dimensione della confezione:
Pezzo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
15815009 1000 Tests Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 15815009 Fornitore BD Biosciences N. del fornitore 566385
  • ONLINE EXCLUSIVE PRICE
    760.00€
    504.90€ / pezzi 1
    Risparmia 255.10€
    33% Sconto
 Articolo soggetto a restrizioni
Spedizione stimata: 07-12-2026
Buffer formulated for the immunofluorescent staining of cells.
  • The BD Horizon Brilliant Stain Buffer Plus is a buffer formulated for the immunofluorescent staining of cells.
  • This buffer was developed to have a reduced test volume for applications where total staining volume is a concern
  • . Brilliant Stain Buffer Plus is as effective as BD Horizon Brilliant Stain Buffer (Cat. No. 563794/566349) in mitigating staining artifacts seen when using multiple BD Horizon Brilliant reagents.
  • For optimal and reproducible results, either BD Horizon Brilliant Stain Buffer Plus or BD Horizon Brilliant Stain Buffer should be used anytime two or more BD Horizon Brilliant™ dye-conjugated reagents are used in the same experiment.
  • The BD Horizon Brilliant Stain Buffer Plus is compatible with the use of other fluorescent staining reagents conjugated with traditional fluorochromes, such as fluorescein, phycoerythrin or Alexa Fluor™ dyes.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Brilliant stain buffer
Quantità 1000 Tests