Learn More
Mettler Toledo™ Bromide Membrane Kit
Product Code. 10340852
Change view
Click to view available options
Material:
Solid State Membrane
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10340852 Solid State Membrane 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10340852 Supplier Mettler Toledo™ Supplier No. 51340006
  • ONLINE EXCLUSIVE PRICE
    715.00€
    607.20€ / 1 sets
    Risparmia 107.80€
    15% Sconto
Spedizione stimata: 01-02-2027
Specifications
For Use With (Application) In gold Bromide ISE with 3 Membrane
Material Solid State Membrane
Temperature (Metric) Operating 0°C to 80°C
pH Range 2 to 13