Learn More
Emma Safety Footwear Bryce Safety Shoes
The impenetrable sole protects your feet from sharp and metal objects while the steel toe cap and reinforced PU toe cover offer protection from falling objects. Microfiber construction remains flexible to stand up to cement and resists corrosive substances.
Marca: Emma Safety Footwear MM33084849
Dettagli aggiuntivi : Peso : 2.16600kg
Descrizione
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo Heat Resistant Outsole
- Microfiber construction
- 3M™ Reflective Striping
- Hydro-Tec™ Sanitized Silver
Specifica
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
49 |
Laces | |
Bryce | |
Microfiber | |
Steel |