Learn More
BTZ043, MedChemExpress
Product Code. 16498437
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16498437 5 mg 5mg
16461917 50 mg 50mg
16436236 100 mg 100mg
16415768 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16498437 Supplier Medchem Express Supplier No. HY13579
 Restricted Item
Spedizione stimata: 09-12-2026
MedChemExpress BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.
BTZ043 is an inhibitor of decaprenyl-phosphoribose-epimerase (DprE1), with MICs of of 2.3 nM and 9.2 nM for M. tuberculosis H37Rv and Mycobacterium smegmatis, respectively.
Specifications
Chemical Name or Material BTZ043
CAS 1161233-85-7
Color Yellow
Purity Grade Notes Research
Molecular Formula C17H16F3N3O5S
Quantity 5 mg
Solubility Information DMSO : 13.3 mg/mL (30.83 mM; Need ultrasonic and warming)
SMILES O=C1N=C(N(CC2)CCC32OC[C@H](C)O3)SC4=C([N+]([O-])=O)C=C(C(F)(F)F)C=C14
Molecular Weight (g/mol) 431.39
Formula Weight 431.39
Percent Purity 99.75%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only