Descrizione
- For use with V700/710 vacuum pumps
- Includes retaining ring for V-700
- May be connected in front of pump as a vacuum reservoir to protect pump from liquids accidentally sucked into vacuum line
- Recommended for stand-alone systems
- GL40, 125 mL capacity
Specifica
Specifica
| Materiale | Borosilicate Glass (3.3) |
| Tipo | Woulff Bottle |
| Da utilizzare con (apparecchiatura) | V-700/710 vacuum pumps |
| Volume (sistema metrico) | 125 mL |