missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, Duracal, pH 10.01 (25°C), Honeywell Fluka™
500 mL, certified buffer solution traceable to NIST/PTB reference materials, DKD certified
Marca: Honeywell Fluka 242148-500ML
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifica
Buffer Solution Duracal Ph 10.01 (25°C) | |
Yellow | |
Poly bottle |
Certified buffer solution traceable to NIST/PTB reference materials, DKD certified | |
500 mL | |
NONH for all modes of transport |