Learn More
Buffer Solution Duracal pH 7.00 (25°C),NIST/PTB reference materials, DKD certified, Honeywell Fluka™
250 mL, certified buffer solution traceable to NIST/PTB reference materials, DKD certified
Marca: Honeywell Fluka 242145-250ML
Dettagli aggiuntivi : Peso : 0.25250kg
Descrizione
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifica
Buffer Solution Duracal Ph 7.00 (25°C) | |
Green | |
NONH for all modes of transport |
Certified buffer solution traceable to NIST/PTB reference materials, DKD certified | |
250 mL |