Learn More
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical™
Sfoglia Tutto Fisher Chemical Prodotti
Click to view available options
Quantità:
500 mL
1 L
2.5 L
Dimensione della confezione:
1L
2.50L
500mL
Descrizione
Buffer Solution pH6.00 (Phosphate), NIST Standard Solution ready to use for pH measurement, Fisher Chemical, Quantity: 1L, CAS: 7778-77-0, Colorless, Packaging: HDPE plastic bottle, pH: 6, MDL Number: 81857, Physical Form: Liquido
Specifica
Specifica
| CAS | 7778-77-0 |
| Colore | Colorless |
| pH | 6 |
| Forma fisica | Liquid |
| Quantità | 500 mL |
| Numero MDL | 81857 |
| Imballaggio | HDPE plastic bottle |