Learn More
Cytiva Buffer Valve
Product Code. 15289764 Shop All Cytiva Products
Change view
Click to view available options
For Use With (Equipment):
AKTA start Automated Protein Purification System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15289764 AKTA start Automated Protein Purification System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15289764 Supplier Cytiva Supplier No. 29023895
In stock
For use with the AKTA start automated protein purification system. The buffer valve is used as a switching valve for gradient formation. It enables the use of two buffers, A and B, which are required for forming gradient during runs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Used as a switching valve for gradient formation
For Use With (Equipment) AKTA start Automated Protein Purification System