Learn More
Kartell™ Burette Clamp
Product Code. 15835567
Change view
Click to view available options
Diameter (Metric) Rod:
8 to 14 mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Rod Unit Size
15835567 8 to 14 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15835567 Supplier Kartell™ Supplier No. 0013900
  • ONLINE EXCLUSIVE PRICE
    93.00€
    83.15€ / pezzi 5
    Risparmia 9.85€
    10% Sconto
In stock
Kartell™ Burette Clamp used a spring loaded center arm to hold the burette firmly against the two outer arms and is protected by rubber grips.
  • Can fit on to support rods having diameters 8 - 14mm
  • Stainless steel spring loaded centre arm holds burette firmly against the two outer arms and is protected by rubber grips
  • Graduations and meniscus are visible at all times
  • Suitable for all burettes
Specifications
Color White
For Use With (Equipment) 8 to 14 mm Rods
Material Polypropylene
Includes 1 Clamp
Type Buret Clamp
Diameter (Metric) Rod 8 to 14 mm
Quantity 1 Ea.