Learn More
Lenz Laborglasintrumente Burette Stopcock, Lateral, with PTFE Plug
Descrizione
- Facetted tip
- Stopcock: NS 12.5
- Bore: 1.5 mm
- NS PTFE plug
- Screwthread retaining device
- Material: borosilicate glass 3.3
Specifica
Specifica
| Materiale | Borosilicate Glass (3.3) |
| Valvola di chiusura | NS 12.5 |