Learn More
Uvex Busingess Gray PU 84699 Low Shoe - Width 12
Uvex 1 shoes designed for classic business look while providing protection against workplace risks; extra-wide steel toecap, penetration-resistant midsole and SRC polyurethane outsole protects toes and keeps from slipping; ideal for active men in office and factory environments
Marca:  Uvex 8469947
Dettagli aggiuntivi : Peso : 1.39000kg
 vedi altre versioni di questo prodotto
Codice prodotto. 17274601
  • 129.00€ / 1 paio
Spedizione stimata: 28-03-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Uvex 1 business Shoe 8469.9 S2 SRC
  • Safe, smart, stylish
  • Fashionably business look with trendy design
  • Generous fit with wide steel toe cap
  • Uvex Climazone technology ensures optimum wearer comfort
  • Breathable, water-resistant high-tech microvelour upper material
  • Textile lining provides optimum breathability
  • Made from synthetic materials, suitable for people allergic to chrome
  • Shock absorbing, durable mono density polyurethane sole unit
  • Removable comfort insole, antistatic with moisture control system and shock absorption in heel and at front of foot
  • Soft padded collar and tongue
  • Sole unit of free of lacquer wetting inhibitors such as silicones, phthalates, etc.
  • Fulfills ESD requirements, resistance to ground Kleiner 35 megohms
  • Certified with Uvex medicare for individual, orthopedic inlay soles and individual adjustments of sole unit such as overall increase, inside and outside increase and other solutions
  • Article No. 8469.9
  • Standard EN ISO 20345:2011 S2 SRC
  • Sizes: 39 to 52
Specifica
Specifica
Gray
47
Steel
EN ISO 20345:2011
Wide width
Yes
S2
Microvelour
Polyurethane (PU)
No
Men's
Uvex 1
No
SRC
Raccomandazioni
Raccomandazioni