Learn More
Butanedioic-13C4 Acid 1-Methyl Ester, TRC
Specifica
Specifica
| Materiale o nome chimico | Butanedioic-13C4 Acid 1-Methyl Ester |
| CAS | 878205-29-9 |
| Formula molecolare | C13C4H8O4 |
| Quantità | 1 mg |
| Sinonimo | Butanedioic-13C4 Acid 1-Methyl Ester, Butanedioic-13C4 Acid Monomethyl Ester, Succinic-13C4 Acid Methyl Ester, Succinic-13C4 Acid Monomethyl Ester, 3-(Methoxycarbonyl)propanoic-13C4 Acid, 3-(Methoxycarbonyl)propionic-13C4 Acid, 3-Carbomethoxypropanoic-13C4 Acid, 4-Methoxy-4-oxobutanoic-13C4 Acid, Methyl Hydrogen Succinate-13C4, Methyl Monosuccinate-13C4, Monomethyl Butanedioate-13C4, Monomethyl Succinate-13C4, NSC 511-13C4 |
| InChI Formula | InChI=1S/C5H8O4/c1-9-5(8)3-2-4(6)7/h2-3H2,1H3,(H,6,7)/i2+1,3+1,4+1,5+1 |
| SMILES | O[13C]([13CH2][13CH2][13C](OC)=O)=O |
| IUPAC Name | 4-methoxy-4-oxo(1,2,3,4-13C4)butanoic acid |
| Molecular Weight (g/mol) | 136.09 |