Learn More
Butoconazole Nitrate, TRC
Specifications
Specifications
| Materiale o nome chimico | Butoconazole Nitrate |
| CAS | 64872-77-1 |
| Formula molecolare | C19 H17 Cl3 N2 S . H N O3 |
| Quantità | 5 g |
| Sinonimo | (+/-)-1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate, 1-[4-(4-Chlorophenyl)-2-[(2,6-dichlorophenyl)thio]butyl]-1H-imidazole mononitrate, (+/-)-Butoconazole nitrate, Butaconazole nitrate, Butoconazole nitrate, Femstat, Gynomyk |
| InChI Formula | InChI=1S/C19H17Cl3N2S.HNO3/c20-15-7-4-14(5-8-15)6-9-16(12-24-11-10-23-13-24)25-19-17(21)2-1-3-18(19)22;2-1(3)4/h1-5,7-8,10-11,13,16H,6,9,12H2;(H,2,3,4) |
| SMILES | O[N+](=O)[O-].Clc1ccc(CCC(Cn2ccnc2)Sc3c(Cl)cccc3Cl)cc1 |
| IUPAC Name | 1-[4-(4-chlorophenyl)-2-(2,6-dichlorophenyl)sulfanylbutyl]imidazole;nitric acid |
| Molecular Weight (g/mol) | 474.79 |
| Peso formulazione | 473.0134 |