Learn More
Butyl 4,4'-bis(tert-butylperoxy)valerate (Technical Grade), TRC
Codice prodotto. 30468964
Cambia vista
Click to view available options
Quantità:
100 mg
Dimensione della confezione:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30468964 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30468964 Fornitore Toronto Research Chemicals N. del fornitore TRCL474890100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Butyl 4,4'-bis(tert-butylperoxy)valerate (Technical Grade)
CAS 995-33-5
Formula molecolare C17H34O6
Quantità 100 mg
Sinonimo Butyl 4,4-bis(tert-butylperoxy)pentanoate, Butyl 4,4-bis(tert-butylperoxy)valerate, Butyl 4,4-di-tert-butylperoxyvalerate, Butyl 4,4'-bis(tert-butylperoxy)valerate
InChI Formula InChI=1S/C17H34O6/c1-9-10-13-19-14(18)11-12-17(8,22-20-15(2,3)4)23-21-16(5,6)7/h9-13H2,1-8H3
SMILES CCCCOC(=O)CCC(C)(OOC(C)(C)C)OOC(C)(C)C
IUPAC Name butyl 4,4-bis(tert-butylperoxy)pentanoate
Molecular Weight (g/mol) 334.45
Peso formulazione 334.2355