Learn More
Butyl 4,4'-bis(tert-butylperoxy)valerate (Technical Grade), TRC
Codice prodotto. 30479655
Cambia vista
Click to view available options
Quantità:
1 g
Dimensione della confezione:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30479655 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 30479655 Fornitore Toronto Research Chemicals N. del fornitore TRCL4748901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Butyl 4,4'-bis(tert-butylperoxy)valerate (Technical Grade)
CAS 995-33-5
Formula molecolare C17H34O6
Quantità 1 g
Sinonimo Butyl 4,4-bis(tert-butylperoxy)pentanoate, Butyl 4,4-bis(tert-butylperoxy)valerate, Butyl 4,4-di-tert-butylperoxyvalerate, Butyl 4,4'-bis(tert-butylperoxy)valerate
InChI Formula InChI=1S/C17H34O6/c1-9-10-13-19-14(18)11-12-17(8,22-20-15(2,3)4)23-21-16(5,6)7/h9-13H2,1-8H3
SMILES CCCCOC(=O)CCC(C)(OOC(C)(C)C)OOC(C)(C)C
IUPAC Name butyl 4,4-bis(tert-butylperoxy)pentanoate
Molecular Weight (g/mol) 334.45
Peso formulazione 334.2355