missing translation for 'onlineSavingsMsg'
Learn More

C19orf10 Recombinant Protein (P01), Abnova

Marca:  Abnova H00056005-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16162023

  • 458.00€ / 25µg


Spedizione stimata
27-11-2020
Aggiungi al carrello

Descrizione

Descrizione

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifica

Specifica

AAH10129
Solution
56005
C19orf10 (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C19orf10
Human
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
44.77
8
Glutathione Sepharose 4 Fast Flow
25 ug
MAAPSGGWNGVGASLWAALLLGAVALRPAEAVSEPTTVAFDVRPGGVVHSFSHNVGPGDKYTCMFTYASQGGTNEQWQMSLGTSEDHQHFTCTIWRPQGKSYLYFTQFKAEVRGAEIEYAMAYSKAAFERESDVPLKTEEFEVTKTAVAHRPGAFKAELSKLVIVAKASRTEL
EUROIMAGE1875335|IL25|IL27|IL27w|R33729_1|SF20
C19orf10
Wheat Germ (in vitro)
GST
Videos
SDS
Documenti