missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C1q Monoclonal Antibody (3R9/2)
GREENER_CHOICE

Codice prodotto. 11564703
Cambia vista
Click to view available options
Quantità:
100 μL
Dimensione della confezione:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
11564703 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 11564703 Fornitore Invitrogen™ N. del fornitore MA183963

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Mouse Monoclonal Antibody

A suggested positive control for immunohistochemical applications for this product is kidney tissue from patients with streptococcal glomerulonephritis. Strong staining was observed in the glomeruli and in some cases the peritubular capillary (potentially clinically significant). Mouse anti human C1q antibody, clone 004-43.X (3R9/2), recognizes human complement component 1 q (C1q), a approximately 156 kDa secreted protein. C1q associates with proenzymes C1r and C1s to form the calcium-dependent C1 complex, the first component of the serum complement system.

C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
TRUSTED_SUSTAINABILITY

Specifica

Antigene C1q
Applicazioni ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry
Classificazione Monoclonal
Clone 3R9/2
Concentrazione 1.1 mg/mL
Coniugato Unconjugated
Formulazione BBS with < 0.1% sodium azide; pH 8.4
Gene C1QA
N. accesso geni P02745, P02746, P02747
Alias gene AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1Q-C; C1QG; Ciqc; complement C1q A chain; complement C1q B chain; complement C1q C chain; complement C1q chain A; complement C1q chain B; complement C1q subcomponent subunit A; complement C1q subcomponent subunit A-like protein; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide; complement component C1q, A chain; complement component C1q, B chain; complement subcomponent C1q chain B
Simboli geni C1QA, C1QB, C1qc
Specie ospite Mouse
Immunogeno Globular head domain of C1q, purified from human plasma.
Metodo di purificazione Protein A
Quantità 100 μL
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 712, 713, 714
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Forma Liquid
Isotype IgG1
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.