Learn More
Merck Millipore Calbiochem™ JAK Inhibitor I
JAK Inhibitor I, CAS 457081-03-7, is a potent, reversible, cell-permeable, and ATP-competitive inhibitor of JAK 1 (IC50 = 15 nM), JAK2 (IC50 = 1 nM), JAK3 (Ki = 5 nM) and Tyk2 (IC50 = 1 nM).
Marca: Merck Millipore 420099-500UG
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
- Packaged Under Inert Gas
- Cell Permeable
Specifica
98% by HPLC | |
Protect from Light | |
murine JAK1 | |
C18H16FN3O | |
DMSO |
Off-white solid | |
500μg | |
457081-03-7 | |
309.3g/mol |