Learn More
R&D Systems™ Calcein AM
Descrizione
- Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
- Do not use past expiration date.
- The hydrolysis of Calcein AM by intracellular esterases produces calcein (structure B), a hydrophilic, strongly fluorescent compound that is well-retained in the cell cytoplasm.
- Cells grown in black-walled plates can be stained and quantified in less than two hours.
- Non-fluorescent, hydrophilic compound that easily permeates intact, live cells
Specifica
Specifica
| Content And Storage | Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Quantità | 50 μg |