Learn More
Abcam Calcein AM, fluorescent dye for cell viability
Codice prodotto. 32116496 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
32116496 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32116496 Fornitore Abcam N. del fornitore ab14142050ug
 Articolo soggetto a restrizioni
Spedizione stimata: 25-11-2026
MW 994.9 Da, Purity >96%. Cell permeant probe used to determine cell viability in most eukaryotic cells. In live cells the nonfluorescent calcein AM is converted to a green-fluorescent calcein after acetoxymethyl ester hydrolysis by intracellular esterases.
MW 994.9 Da, Purity >96%. Cell permeant probe used to determine cell viability in most eukaryotic cells. In live cells the nonfluorescent calcein AM is converted to a green-fluorescent calcein after acetoxymethyl ester hydrolysis by intracellular esterases.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 50 μg
Status giuridico RUO