Learn More
Thermo Scientific™ Calcium Reagents, Infinity Calcium Arsenazo III 2 x 125mL
Codice prodotto. 17882041 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
2 x 125 mL
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
17882041 2 x 125 mL pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17882041 Fornitore Thermo Scientific™ N. del fornitore 1265250
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Visualizzare prodotti alternativi
Determine in quantity of calcium in serum or plasma in vitro with Thermo Scientific™ Calcium Reagents.
Calcium plays an essential role in bone mineralization, blood coagulation, muscle contraction and relaxation, nerve signaling and release of certain hormones. Increased levels are associated with hyperparathyroidism, metastatic bone lesions, and Vitamin A or D overdose. Decreased levels are most commonly associated with hypoalbuminemia but can also be observed in cases of hypoparathyroidism, Vitamin D deficiency or resistance, chronic renal failure and calcium losing syndromes. Reagent has up to 18 months of stability from the date of manufacture.
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità CE Marked
Content And Storage 2°C to 8°C
Descrizione Infinity Calcium Arsenazo III
Marchio CE Yes
Analiti rilevabili Calcium
Tipo di campione Plasma, Serum
Tipo Reagent
Linearità 6 to 15mg/dL
Compatibilità metodo Endpoint
Quantità 2 x 125 mL
Stabilità Stable Liquid
Stato a temperatura ambiente Liquid
Forma Liquid