Learn More
Endecotts™ Sieve Calibration Standards
Descrizione
- Calibrates sieves to an accuracy of ±1μm
- Soda-lime glass microspheres pass over the entire surface of the sieve and through every aperture.
- Traceable to the United Kingdom National Physical Laboratory
- Standards include a calibration chart.
Specifica
Specifica
| Product Type | Calibration Sample |
| Materiale | Soda-lime glass |
| Da utilizzare con (apparecchiatura) | Sieve |
| Quantità confezione | 5/Pk. |
| Dimensione particelle | 250 μm |