Learn More
Draeger Calibration Gas, H2S, CO, CO2, CH4, O2/N2
Product Code. 15330448
Change view
Click to view available options
Volume (Metric) Gas:
58 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Gas Unit Size
15330448 58 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15330448 Supplier Draeger Supplier No. 6811132
  • ONLINE EXCLUSIVE PRICE
    883.95€
    539.22€ / pezzi 1
    Risparmia 344.73€
    38% Sconto
 Restricted Item
Spedizione stimata: 21-12-2026
Test gas in disposable bottles. Control valve not supplied (please enquire).
  • 58L disposable bottle
  • According to ISO 9002.DOT39 and EN 12205
Specifications
Certifications/Compliance ISO 9002.DOT39 and EN 12205
Concentration 2.5 Vol.%: 15 ppm, 2 Vol.%: 50 ppm
Gas Type H2S, CO, CO2
Volume (Metric) Gas 58 L
Pressure 35 bar