missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calnexin Mouse, Clone: CANX/1541, enQuireBio™
Mouse Monoclonal Antibody
Marca: EnquireBio 821-MSM1-P1
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Calnexin | |
Cytoplasmic | |
CANX/1541 | |
Unconjugated | |
NP_001737 | |
Mouse | |
90kDa | |
Protein A/G Purified | |
RUO | |
821 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
CANX | |
Recombinant N-terminal fragment of human Calnexin protein | |
HeLa, MCF-7 or U2OS cells. Kidney or small intestine. | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |