missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calponin Mouse anti-Human, Clone: 1A11E5, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Specifica
Antigene | Calponin |
---|---|
Clone | 1A11E5 |
Concentrazione | 1 mg/mL |
Applicazioni | Western Blot, Immunohistochemistry (Paraffin) |
Classificazione | Monoclonal |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16808515
|
Proteintech
66540-1-IG-20UL |
20 μL |
140.00€
20µL |
Spedizione stimata: 14-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16898505
|
Proteintech
66540-1-IG-150UL |
150 μL |
399.00€
150µL |
Spedizione stimata: 14-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
This antibody raised against the specific region of human CNN1, thus it does not cross-react with CNN2 or CNN3.
Calponin, a thin filament associated protein is implicated in the regulation and modulation of smooth muscle contraction. It is capable of binding to actin, calmodulin, troponin C and tropomyosin. Calponin is expressed in smooth muscle and tissues containing significant amounts of smooth muscle. Two isoforms of calponin exist whose molecular weights are 34kDa and 29kDa. Expression of the 29kDa form is primarily restricted to muscle of the urogenital tract. The expression of calponin has also been demonstrated in myoepithelial cells from benign and maligt breast lesions. It stains smooth muscle, myoepithelial cells, myofibroblasts, keratinocytes and nerve fibers. It identifies myoepithelial cells in breast lesions, and helps differentiate breast collagenous spherulosis (positive) from adenoid cystic carcinoma. Adenoid cystic carcinoma in salivary gland tumors is calponin positive.Specifica
Calponin | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Basic calponin, Calponin 1, Calponin H1, smooth muscle, CNN1, Sm Calp, SMCC | |
CNN1 | |
IgG2b | |
Protein A | |
Antibody |
1A11E5 | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
P51911 | |
1264 | |
Calponin Fusion Protein Ag21384 | |
Primary | |
-20°C | |
CNN1 |