missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calumenin Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 17804-1-AP-20UL
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Caluminin is a 315 amino acid Ca2+-binding member of the CREC, EF-hand protein family. Calumenin is a secreted protein t hat contains six Ca2+-binding (EF-hand) motifs and is expressed in the lumen of the endoplasmic reticulum (ER) and Golgi apparatus. In the presence of Ca2+, Calumenin interacts with serum amyloid P component (SAP) and, together, they may play a role in the immunological defense system and participate in amyloidosis, the pathological formation of amyloid deposits in different types of tissues. Calumenin has an inhibitory effect on the vitamin K-dependent g-carboxylation system which converts vitamin K-dependent proteins to Gla-containing proteins. Calumenin may also be involved in the pathophysiology of thrombosis and/or wound healing by acting in an autocrine or paracrine manner.Specifica
Calumenin | |
Polyclonal | |
Unconjugated | |
CALU | |
CALU, Calumenin, Crocalbin, IEF SSP 9302 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
12321, 813 | |
-20°C | |
Liquid |
Immunofluorescence, Immunocytochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35887, O43852 | |
CALU | |
Calumenin Fusion Protein Ag12073 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |