Learn More
Motic™ Camera Adapter, C-Mount 0.5X, 1in.
Product Code. 15405112
Change view
Click to view available options
Magnification Power:
0.5X
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Magnification Power Unit Size
15405112 0.5X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15405112 Supplier Motic™ Supplier No. 1101005600031
  • ONLINE EXCLUSIVE PRICE
    75.00€
    66.64€ / pezzi 1
    Risparmia 8.36€
    11% Sconto
Spedizione stimata: 25-01-2027
Choose the most appropriate adapter for your camera and microscope and unlock the potential of photomicography. Motic™ Camera Adapter, C-Mount 0.5X, 1in. uses the industry standard attachment for digital imaging devices dedicated to microscopes.
  • Connect a 1-inch sensor C-mount camera with this adapter.
  • Suitable for Series 28 Educational Microscopes.
Specifications
Type C-mount Adapter
Connector Types C-Mount
For Use With (Equipment) Series 28 Educational Microscopes, 1in. Sensor C-Mount Camera
Magnification Power 0.5X