missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX cAMP Hunter™ CHO-K1 HTR7A Gs Cell Line

Codice prodotto. 17304689
Encompass
Cambia vista
Click to view available options
Quantità:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
17304689 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17304689 Fornitore DiscoverX N. del fornitore 950151C2

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The cAMP Hunter™ CHO-K1 HTR7A Gs Cell Line measures HTR7A (GPCR) activity via increased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.

The cAMP Hunter HTR7A Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin/5-HT) based activation of HTR7A GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype HTR7A, a naturally Gs coupled GPCR. Upon activation of HTR7A, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifica

Assay Entry Intracellular cAMP level
Linea cellulare CHO-K1
Da utilizzare con (applicazione) Cell Lines
Primary Agonist Serotonin/5-HT
Quantità 2 Vials
Target Entry HTR7A
Numero di accesso NM_000872.4
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.