missing translation for 'onlineSavingsMsg'
Learn More

DiscoverX cAMP Hunter™ CHO-K1 SSTR4 Gi Cell Line

Codice prodotto. 17334489
Encompass
Cambia vista
Click to view available options
Quantità:
2 Vials
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
17334489 2 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 17334489 Fornitore DiscoverX N. del fornitore 950059C2

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

The cAMP Hunter™ CHO-K1 SSTR4 Gi Cell Line measures SSTR4 (GPCR) activity via decreased levels of intracellular cAMP. This cell line ships as two vials of cryopreserved cells.

The cAMP Hunter SSTR4 Gi cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. somatostatin 28) based activation of SSTR4 GPCR activity via detection of decreasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wildtype SSTR4 , a naturally Gi/Go coupled GPCR. Upon activation of SSTR4 , the decrease in levels of intracellular cAMP leads to a decreased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells.

Specifica

Assay Entry Intracellular cAMP level
Linea cellulare CHO-K1
Da utilizzare con (applicazione) Cell Lines
Primary Agonist Somatostatin 28
Quantità 2 Vials
Target Entry SSTR4
Numero di accesso AY548169
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.