Learn More
Cancer-Targeting Compound 1, MedChemExpress
Product Code. 16483309
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16483309 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16483309 Supplier Medchem Express Supplier No. HYU00300
 Restricted Item
Spedizione stimata: 09-12-2026
MedChemExpress Cancer-Targeting Compound 1 is used in the research of hormone-​related cancer, extracted from patent WO 2008021331 A2.
Cancer-Targeting Compound 1 is used in the research of hormone-​related cancer, extracted from patent WO 2008021331 A2.
Specifications
Chemical Name or Material Cancer-Targeting Compound 1
CAS 1007581-62-5
Color White
Purity Grade Notes Research
Molecular Formula C15H13N3O2S
Quantity 1 mg
SMILES N#CC1=CC=C(C2=CSC(N3C(OCC3(C)C)=O)=N2)C=C1
Molecular Weight (g/mol) 299.35
Formula Weight 299.35
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less
Research purposes only