Learn More
Thermo Scientific™ Thermo Scientific™ Hair/Nail Specimen Collection Kit
Description
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kits enable convenient collection and transport of samples for drugs of abuse or forensic testing. Each kit contains a collection foil, transport envelope, specimen bag, and tamper-evident tape.
Features:
- Tamper-evident closures
- Collection containers are Class 10,000 aseptic products
- Customized kits and OEM parts available
- Leak-resistant & air-tight, with a lock seal
- Twin valve design provides tighter seal
Specifications
Specifications
| Product Type | Specimen Collection Kit |
| Descrizione | Hair / Nail Specimen Collection Kit, with foil, envelope, seal tape |
| Include | Foil, Envelope, Seal Tape |
| N. per custodia | 100/Cs. |