Learn More
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kit
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kits enable convenient collection and transport of samples for drugs of abuse or forensic testing.
Marca: Thermo Scientific™ 98047
Dettagli aggiuntivi : Peso : 0.95000kg
Descrizione
Thermo Scientific™ Capitol Vial Hair/Nail Specimen Collection Kits enable convenient collection and transport of samples for drugs of abuse or forensic testing. Each kit contains a collection foil, transport envelope, specimen bag, and tamper-evident tape.
Features:
- Tamper-evident closures
- Collection containers are Class 10,000 aseptic products
- Customized kits and OEM parts available
- Leak-resistant & air-tight, with a lock seal
- Twin valve design provides tighter seal
Specifica
Specimen Collection Kit | |
Hair / Nail Specimen Collection Kit, with foil, envelope, seal tape | |
100/Cs. |