Learn More
Erlab™ Captair 834 Smart Filtering Storage Cabinet
Erlab™ Captair 834 Smart Filtering Storage Cabinets provide a steady flow of air through the cabinet, capturing hazardous vapors in the filters, and returns clean air back to the room.
dietro preventivo
Specifica
Certificazione/Conformità | Filtration performances tested according to the AFNOR NF X 15-211:2009 standard : France EN 1822 : 1998 (HEPA H14 & ULPA U17 Filters) - CE Marking |
---|---|
Profondità (sistema metrico) | 500 mm |
Descrizione | Power Consumption: 45 W |
Product Type | Ductless Filtering Storage Cabinet |
Altezza (sistema metrico) | 2140 to 2300 mm |
Descrizione
- Filter AS - For organic vapors
- Filter BE - For acid vapors
- Filter F - For formaldehyde vapors
- Type 1C includes one carbon filter
- Type 1P2C includes one particulate filter and one two carbon filters
- Air Flow: 220 m3/h / 129 CFM
- Electrical Requirements: 80 to 220 V, 50/60 Hz
- Corrosion resistant electro-galvanized steel coated with anti-acid polymer
- Doors: Clear, chemical resistant acrylic for easy viewing
- 1 column that can be configured to handle liquids, powders, or both
- eGuard app (Android or iOS): For remote control to monitor the status of the storage cabinet, change the settings, and deliver safety alerts immediately to your devices (mobile, tablet and PC)
- Alarm if doors are left open
Specifica
Filtration performances tested according to the AFNOR NF X 15-211:2009 standard : France EN 1822 : 1998 (HEPA H14 & ULPA U17 Filters) - CE Marking | |
Power Consumption: 45 W | |
2140 to 2300 mm | |
Up to 120 x 1 L Bottles | |
5 |
500 mm | |
Ductless Filtering Storage Cabinet | |
100 to 120 bottles | |
2 | |
900 mm |