Learn More
Thermo Scientific™ CaptureSelect™ AAVX Ligand Leakage ELISA
Codice prodotto. p-7180723 Sfoglia Tutto Thermo Scientific Prodotti
Cambia vista
Click to view available options
Quantità:
1 Assay
10 Assays
Dimensione della confezione:
Pezzo
pezzi 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
30067389 1 Assay Pezzo
18476408 10 Assays pezzi 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 30067389 Fornitore Thermo Scientific™ N. del fornitore 810352201
Thermo Scientific™ CaptureSelect™ AAVX Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Matrix.
CaptureSelect AAVX Ligand Leakage ELISA is designed to detect possible leached ligand of adeno-associated viruses (AAV) purified using POROS CaptureSelect AAVX Affinity Matrix. The ligand, a 13-kD recombinant protein, is covalently coupled for high chemical stability and low leakage. Even when covalently attached, small amounts of the affinity ligand can leach off of the chromatography support and co-elute with the purified AAV.
This ELISA provides typical sensitivity and accurate quantitation of possible leached ligand greater than or equal to 1 ng/mL. POROS CaptureSelect AAVX Ligand Leakage ELISA can be used as a tool to aid in optimal purification process development and in routine quality control of in-process streams as well as final product.
Specifica
Content And Storage 3 x 0.1 mL, store at -5°C to -30°C.
Linea di prodotti CaptureSelect
Quantità 1 Assay
Shipping Condition Dry Ice