Learn More
Carbadox, TRC
Product Code. 30007207
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007207 5 g 5g
30040566 100 mg 100mg
30007647 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30007207 Supplier Toronto Research Chemicals Supplier No. TRCC1758255G
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Identificatori chimici
CAS 6804-07-5
Molecular Formula C11 H10 N4 O4
Molecular Weight (g/mol) 262.22
Synonym Carbazic acid, 3-(2-quinoxalinylmethylene)-, methyl ester, N,N'-dioxide (7CI), Carbazic acid, 3-(2-quinoxalinylmethylene)-, methyl ester, N1,N4-dioxide (8CI), Hydrazinecarboxylic acid, (2-quinoxalinylmethylene)-, methyl ester, N,N'-dioxide, Hydrazinecarboxylic acid, [(1,4-dioxido-2-quinoxalinyl)methylene]-, methyl ester (9CI), 2-Formylquinoxaline-1,4-dioxide carbomethoxyhydrazone, Carbadox, Fortigro, GS 6244, Getroxel, Mecadox, Methyl 3-(2-quinoxalinylmethylene)carbazate N1,N4-dioxide, Quinoxaline-2-carbomethoxyhydrazone 1,4-dioxide
IUPAC Name methyl N-[(E)-(1,4-dioxidoquinoxaline-1,4-diium-2-yl)methylideneamino]carbamate
SMILES COC(=O)N\N=C\c1c[n+]([O-])c2ccccc2[n+]1[O-]
Specifica
Quantity 5 g
InChI Formula InChI=1S/C11H10N4O4/c1-19-11(16)13-12-6-8-7-14(17)9-4-2-3-5-10(9)15(8)18/h2-7H,1H3,(H,13,16)/b12-6+
Formula Weight 262.0702
Percent Purity >95%
Chemical Name or Material Carbadox
Grade HPLC