Learn More
Brannan™ Carbon Dioxide Detector
Product Code. 17435313
Change view
Click to view available options
Components Measured:
Carbon Dioxide
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Components Measured Unit Size
17435313 Carbon Dioxide Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17435313 Supplier Brannan™ Supplier No. 38/920/0
  • ONLINE EXCLUSIVE PRICE
    219.00€
    210.87€ / pezzi 1
    Risparmia 8.13€
    3% Sconto
Spedizione stimata: 04-01-2027
The Brannan carbon dioxide detector measures Carbon Dioxide (CO2), air temperature and humidity. Comes complete with certificate of calibration.
  • Measures ventilation in enclosed spaces such as classrooms and workplaces
  • Indoor air quality displayed in ppm with colour coded good, normal and poor indicators
  • This device adopts a high-precision non-dispersive infrared CO2 sensor that detects CO2 accurately and sensitively
  • This product uses a high-capacity rechargeable lithium battery inside, making it ideal to move around a space
  • To preserve electricity, the device powers off after 30 minutes of inactivity
  • A USB charging cable is provided allowing the user to recharge the device using either a PC or mains plug
Specifications
Certifications/Compliance CE/RoHS/WEEE
Detection Limits 0 to 9999 ppm, -20°C to +70°C, 0 to 100% RH
Components Measured Carbon Dioxide
Sensor Types Non-dispersive infrared CO2 sensor
Weight (English) 0.5302 lb.
Weight (Metric) 0.2405 kg