Learn More
Carbon Steel, AISI 1045, G10450, 100g, ARMI∣MBH
Codice prodotto. 17969094
Cambia vista
Click to view available options
Quantità:
100 g
Dimensione della confezione:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
17969094 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 17969094 Fornitore ARMI/MBH N. del fornitore IARM200DC
Please to purchase this item. Need a web account? Register with us today!
ARMI∣MBH produces high-quality metal alloy certified reference materials and set up standards for Spark-OES, ICP, and XRF elemental analysis. The offering includes steel, aluminum, titanium, nickel, cobalt, copper, magnesium, and more.
Carbon Steel, AISI 1045, G10450, 100g
TRUSTED_SUSTAINABILITY
Specifica
Materiale o nome chimico Titanium/Niobium/Tungsten/Aluminium/Arsenic/Cobalt/Tin/Nitrogen/Phosphorus/Molybdenum/Vanadium/Nickel/Chromium/Silicon/Copper/Carbon/Manganese/Magnesium/Lead/Tantalum/Boron/Calcium/Antimony/Zinc/Oxygen
CAS 7439-89-6
Quantità 100 g
Forma Pieces
Concentration or Composition (by Analyte or Components) Titanium 0.00013 %/Niobium 0.001 %/Tungsten 0.003 %/Aluminium 0.004 %/Arsenic 0.005 %/Cobalt 0.007 %/Tin 0.0079 %/Nitrogen 0.009 %/Phosphorus 0.0103 %/Molybdenum 0.0217 %/Vanadium 0.0244 %/Nickel 0.097 %/Chromium 0.109 %/Silicon 0.225 %/Copper 0.232 %/Carbon 0.453 %/Manganese 0.749 %/Magnesium (<0.001) %/Lead (<0.002) %/Tantalum (<0.005) %/Boron (0.0002) %/Calcium (0.001) %/Antimony (0.0019) %/Zinc (0.002) %/Oxygen (0.004) %