Learn More
Thermo Scientific Chemicals Carboxymethylecellulose, sodium salt, average M.W. 250000 (DS=0.9)
Click to view available options
Quantità:
100 g
1 kg
Imballaggio:
Glass bottle
Plastic bottle
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Materiale o nome chimico | Carboxymethylecellulose, sodium salt |
Colore | White to Beige |
Nota nome | Average M.W. 250000 (DS=0.9) |
pH | 6.5 to 8 (1% soln.) |
Forma fisica | Crystalline Powder |
Indice di Merck | 15, 1830 |
Quantità | 100 g |
Percent Purity | ≥99.5% |
Spettro a infrarossi | Authentic |
Perdita all'essiccatura | 10% max. (as packed) (3 to 5 g, 105°C, 2 hrs) |